missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTHFR Polyclonal Antibody
MTHFR Polyclonal Antibody
Brand: Invitrogen PA5142473
This item is not returnable.
View return policy
Description
MTHFR Polyclonal Antibody for IHC (P)
Specifications
| MTHFR | |
| Polyclonal | |
| Unconjugated | |
| MTHFR | |
| 5,10-methylenetetrahydrofolate reductase; 5,10-methylenetetrahydrofolate reductase (NADPH); AI323986; Methylenetetrahydrofolate reductase; methylenetetrahydrofolate reductase (NAD(P)H); MTHFR; RP11-56N19.4 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 4524 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P42898 | |
| MTHFR | |
| Peptide with sequence C-NRPTQNARETEAP (aa 644-656). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction