Learn More
MTHFR Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26591-1-AP-150UL
This item is not returnable.
View return policy
Description
The MTHFR antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MTHFR. This antibody recognizes human antigen. The MTHFR antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| MTHFR | |
| 250 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| MTHFR | |
| 150 μL | |
| 4524 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |