Learn More
MTHFR Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26591-1-AP-20UL
 View more versions of this product
Product Code. 18095485
  • € 145.00 / 20µL
Description
The MTHFR antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MTHFR. This antibody recognizes human antigen. The MTHFR antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
MTHFR
250 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, ELISA
MTHFR
20 μL
4524
Store at -20°C. Stable for one year after shipment.