missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTIF3 Monoclonal Antibody (OTI4B6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526442
This item is not returnable.
View return policy
Description
MTIF3 Monoclonal Antibody for Western Blot, IHC (P)
This gene encodes a translation initiation factor that is involved in mitochondrial protein synthesis. Polymorphism in this gene is associated with the onset of Parkinson's disease. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 5.
Specifications
| MTIF3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9H2K0 | |
| MTIF3 | |
| Human recombinant protein fragment corresponding to amino acids 56-278 of MTIF3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Canine, Monkey | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4B6 | |
| Unconjugated | |
| MTIF3 | |
| 2810012L14Rik; AI414549; DC38; IF3(mt); IF-3(Mt); IF3mt; IF-3Mt; mitochondrial translational initiation factor 3; Mtif3; Translation initiation factor IF-3, mitochondrial | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 219402, 477328 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction