missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTIF3 Monoclonal Antibody (OTI4B6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800436
This item is not returnable.
View return policy
Description
This gene encodes a translation initiation factor that is involved in mitochondrial protein synthesis. Polymorphism in this gene is associated with the onset of Parkinson's disease. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 5.Specifications
| MTIF3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9H2K0 | |
| MTIF3 | |
| Human recombit protein fragment corresponding to amino acids 56-278 of human MTIF3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Canine, Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4B6 | |
| Unconjugated | |
| MTIF3 | |
| IF3mt | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 219402, 477328 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction