Learn More
PELI1 Monoclonal Antibody (OTI3A9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA807607
Encompass_Preferred
 View more versions of this product
Product Code. 17578223
  • € 540.87 / 100µL
Description
The Pellino proteins are a highly homologous family of E3 ubiquitin ligases that act as upstream mediators in Toll-like receptor (TLR) pathways that lead to activation of MAP kinases and transcription factors. Pellino 1 is required for interleukin-1-mediated signaling through its interaction with the IRAK4-IRAK-TRAF6 complex, ultimately resulting in the activation of NF-Kappa-B. Like other members of the Pellino family, Pellino 1 is an E3 ubiquitin ligase, able to catalyze the polyubiquitination of IRAK1. It is activated via phosphorylation by either IRAK1 and IRAK4 or the IKK-related kinases IKK-epsilon; and TBK1. In addition to phosphorylation, Pellino 1 activity is also modulated via ubiquitination and sumoylation.
Specifications
Specifications
PELI1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q96FA3
PELI1
Human recombit protein fragment corresponding to amino acids 1-245 of human PELI1 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI3A9
Unconjugated
PELI1
pellino E3 ubiquitin protein ligase 1
Mouse
Affinity Chromatography
RUO
57162
-20° C, Avoid Freeze/Thaw Cycles