Learn More
MTIF3 Mouse anti-Canine, Human, Clone: OTI4B6, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF800436
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a translation initiation factor that is involved in mitochondrial protein synthesis. Polymorphism in this gene is associated with the onset of Parkinson's disease. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 5.Specifications
| MTIF3 | |
| Monoclonal | |
| Unconjugated | |
| MTIF3 | |
| IF3mt | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 219402, 477328 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4B6 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9H2K0 | |
| MTIF3 | |
| Human recombit protein fragment corresponding to amino acids 56-278 of human MTIF3 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Canine, Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.