missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTMR1 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-44289JF646
This item is not returnable.
View return policy
Description
MTMR1 Polyclonal antibody specifically detects MTMR1 in Human samples. It is validated for Western Blot
Specifications
| MTMR1 | |
| Polyclonal | |
| Western Blot | |
| EC 3.1.3, EC 3.1.3.-, EC 3.1.3.48, myotubularin related protein 1, myotubularin-related protein 1 | |
| The epitope recognized by this antibody maps to a region between residue 615 to 665 of human Myotubularin-related protein 1 using the numbering given in entry Q13613.4 (GeneID 8776). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 8776 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction