missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTMR1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-44289
This item is not returnable.
View return policy
Description
Specifically detects MTMR1 in Human samples, and it is validated for Immunoprecipitation, Western Blotting
MTMR1 Polyclonal specifically detects MTMR1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| MTMR1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| MTMR1 | |
| The epitope recognized by this antibody maps to a region between residue 615 to 665 of human Myotubularin-related protein 1 using the numbering given in entry Q13613.4 (GeneID 8776). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg lysate | |
| EC 3.1.3, EC 3.1.3.-, EC 3.1.3.48, myotubularin related protein 1, myotubularin-related protein 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 8776 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction