missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTMR10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570833
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Guinea Pig: 79%; Horse: 77%; Human: 100%; Mouse: 77%.
MTMR10 is a protein coding gene.
Specifications
| MTMR10 | |
| Polyclonal | |
| Unconjugated | |
| MTMR10 | |
| BB128963; C79734; Inactive phosphatidylinositol 3-phosphatase 10; Mtmr10; myotubularin related protein 10; myotubularin-related protein 10 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 54893 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NXD2 | |
| MTMR10 | |
| synthetic peptide directed towards the C-terminal region of human MTMR10. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction