missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTMR12 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97310C
This item is not returnable.
View return policy
Description
MTMR12 Polyclonal antibody specifically detects MTMR12 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MTMR12 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 650 | |
| 3-PAP3PAP, 3-phosphatase adapter subunit, KIAA1682FLJ20476, myotubularin related protein 12, myotubularin-related protein 12,3-phosphatase adapter protein, Phosphatidylinositol-3 phosphate 3-phosphatase adapter subunit, phosphatidylinositol-3 phosphate 3-phosphatase adaptor subunit, phosphatidylinositol-3-phosphate associated protein, PIP3AP | |
| Produced in rabbits immunized with E. coli-derived Human MTMR12 fragment. | |
| 0.1 mL | |
| Protein Phosphatase | |
| 54545 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction