missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTMR14 Antibody (OTI6B6), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72808MFV500
This item is not returnable.
View return policy
Description
MTMR14 Monoclonal antibody specifically detects MTMR14 in Human, Mouse, Rat, Canine samples. It is validated for Flow Cytometry
Specifications
| MTMR14 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 64419 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| OTI6B6 | |
| Flow Cytometry | |
| C3orf29FLJ11546, chromosome 3 open reading frame 29, EC 3.1.3.-, egg-derived tyrosine phosphatase homolog, FLJ22405, FLJ46453, FLJ90311, HCV NS5A-transactivated protein 4 splice variant A-binding protein 1, hEDTP, HJUMPY, jumpy, myotubularin related protein 14, myotubularin-related protein 14, NS5ATP4ABP1 | |
| Full length human recombinant protein of human MTMR14 (NP_001070993) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction