missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTMR3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556962
This item is not returnable.
View return policy
Description
Immunogen sequence: SLNEKWQEHR RSLELSSLAG PGEDPLSADS LGKPTRVPGG AELSVAAGVA EGQMENILQE ATKEESGVEE PAHRAGIEIQ EGKEDPLLEK ESRRKTPEAS AIGLH Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 66%.
This gene encodes a member of the myotubularin dual specificity protein phosphatase gene family. The encoded protein is structurally similar to myotubularin but in addition contains a FYVE domain and an N-terminal PH-GRAM domain. The protein can self-associate and also form heteromers with another myotubularin related protein. The protein binds to phosphoinositide lipids through the PH-GRAM domain, and can hydrolyze phosphatidylinositol(3)-phosphate and phosphatidylinositol(3,5)-bisphosphate in vitro. The encoded protein has been observed to have a perinuclear, possibly membrane-bound, distribution in cells, but it has also been found free in the cytoplasm. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| MTMR3 | |
| Polyclonal | |
| Unconjugated | |
| MTMR3 | |
| 1700092A20Rik; AI255150; AW557713; FYVE (Fab1 YGLO23 Vsp27 EEA1 domain) dual-specificity protein phosphatase; FYVE domain-containing dual specificity protein phosphatase 1; FYVE-DSP1; KIAA0371; mKIAA0371; MTMR3; myotubularin related protein 3; myotubularin-related protein 3; Phosphatidylinositol-3,5-bisphosphate 3-phosphatase; phosphatidylinositol-3-phosphate phosphatase; ZFYVE10; zinc finger FYVE domain-containing protein 10; zinc finger, FYVE domain containing 10 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8897 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13615 | |
| MTMR3 | |
| Recombinant protein corresponding to Human MTMR3. Recombinant protein control fragment (Product #RP-95358). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction