missing translation for 'onlineSavingsMsg'
Learn More

MTMR4 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 188.00 - € 470.00

Specifications

Antigen MTMR4
Dilution Western Blot 1:200-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18647210
View Documents
Novus Biologicals
NBP2-94706-0.02ml
0.02 mL
€ 188.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18677290
View Documents
Novus Biologicals
NBP2-94706-0.1ml
0.1 mL
€ 470.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MTMR4 Polyclonal antibody specifically detects MTMR4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

MTMR4
Western Blot, Immunofluorescence
Unconjugated
Rabbit
Signal Transduction
PBS (pH 7.3), 50% glycerol
9110
IgG
Affinity purified
Western Blot 1:200-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200
Polyclonal
Purified
RUO
Human, Mouse, Rat
EC 3.1.3.48, FYVE-DSP2, KIAA0647zinc finger, FYVE domain containing 11, myotubularin related protein 4, ZFYVE11FYVE domain-containing dual specificity protein phosphatase 2, Zinc finger FYVE domain-containing protein 11
Recombinant fusion protein containing a sequence corresponding to amino acids 920-1195 of human MTMR4 (NP_004678.3). GEATPRRLLSYGCCSKRPNSKQMRATGPCFGGQWAQREGVKSPVCSSHSNGHCTGPGGKNQMWLSSHPKQVSSTKPVPLNCPSPVPPLYLDDDGLPFPTDVIQHRLRQIEAGYKQEVEQLRRQVRELQMRLDIRHCCAPPAEPPMDYEDDFTCLKESDGSDTEDFGSDHSEDCLSEASWEPVDKKETEVTRWVPDHMASHCYNCDCEFWLAKRRHHCRNCGNVFCAGCCHLKLPIPDQQLYDPVLVCNSCYEHIQVSRARELMSQQLKKPIATASS
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.