Learn More
MTMR4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13488-1-AP-150UL
 View more versions of this product
Product Code. 16851724
  • € 410.00 / 150µL
Description
MTMR, a member of the myotubularin family, encodes an intracellular protein of 1, 195 amino acids, and shares approximately 47% sequence identity with MTMR3, spiking to over 70% identity in the catalytic domain and the double zinc finger (FYVE) motif. PCR analysis detects expression of MTMR4 in all human tissues examined except lung, small intestine, stomach, salivary gland, adrenal gland, and uterus. Subcellular localization of MTMR4 in transfected HeLa cells showed a perinuclear distribution, similar to that seen with MTMR3. MTMR4 dephosphorylates para-nitrophenylphosphate and phosphatidylinositol 3-phosphate.
Specifications
Specifications
MTMR4
Polyclonal
Unconjugated
MTMR4
FYVE DSP2, KIAA0647, MTMR4, myotubularin related protein 4, ZFYVE11
Rabbit
Antigen Affinity Chromatography
RUO
170749, 287607, 9110
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q91XS1, Q9NYA4
Mtmr4
MTMR4 Fusion Protein Ag4310
150 μL
Primary
Human, Mouse, Rat