missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTMR6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570556
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Dog: 93%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Rabbit: 86%; Rat: 79%.
Phosphatase that acts on lipids with a phosphoinositol headgroup (PubMed:19038970, PubMed:22647598). Dephosphorylates phosphatidylinositol 3-phosphate (PtdIns(3)P) and phosphatidylinositol 3,5-bisphosphate (PubMed:19038970, PubMed:22647598) (Probable). Binds with high affinity to phosphatidylinositol 3,5-bisphosphate (PtdIns(3,5)P2) but also to phosphatidylinositol 3-phosphate (PtdIns(3)P), phosphatidylinositol 4-phosphate (PtdIns(4)P), and phosphatidylinositol 5-phosphate (PtdIns(5)P), phosphatidic acid and phosphatidylserine (PubMed:19038970). Negatively regulates ER-Golgi protein transport. Probably in association with MTMR9, plays a role in the late stages of macropinocytosis by dephosphorylating phosphatidylinositol 3-phosphate in membrane ruffles (PubMed:24591580). Acts as a negative regulator of KCNN4/KCa3.1 channel activity in CD4(+) T-cells possibly by decreasing intracellular levels of phosphatidylinositol 3-phosphate (PubMed:15831468). Negatively regulates proliferation of reactivated CD4(+) T-cells (PubMed:16847315). In complex with MTMR9, negatively regulates DNA damage-induced apoptosis (PubMed:19038970, PubMed:22647598). The formation of the MTMR6-MTMR9 complex stabilizes both MTMR6 and MTMR9 protein levels (PubMed:19038970). [UniProt].
Specifications
| MTMR6 | |
| Polyclonal | |
| Unconjugated | |
| MTMR6 | |
| 4022440C11Rik; AI428804; AU041072; MTMR6; myotubularin related protein 6; Myotubularin-like protein 6-like protein; myotubularin-related protein 6; Phosphatidylinositol-3,5-bisphosphate 3-phosphatase; Phosphatidylinositol-3-phosphate phosphatase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 9107 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y217 | |
| MTMR6 | |
| synthetic peptide directed towards the middle region of human MTMR6 (aa 531-580). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction