Learn More
MTOR Monoclonal Antibody (OTI1G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA805901
Encompass_Preferred
 View more versions of this product
Product Code. 17535923
  • € 540.87 / 100µL
Description
Mammalian target of rapamycin (mTOR) is a serine/threonine kinase that plays a key role in cell growth, cell proliferation, and protein synthesis. mTOR mediates phosphoinositide 3-kinase and Akt/PKB signaling, resulting in phosphorylation of 4EBP1, and initiation of mRNA translation. A second pathway involves regulation of ribosomal S6 kinase, which affects ribosome biogenesis and translation elongation. Mutations affecting the gene results in Smith-Kingsmore syndrome.
Specifications
Specifications
MTOR
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P42345
MTOR
Human recombit protein fragment corresponding to amino acids 1766-2144 of human MTOR produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2a
Western Blot
OTI1G4
Unconjugated
MTOR
FRAP, FRAP1, FRAP2, RAFT1, RAPT1, SKS
Mouse
Affinity Chromatography
RUO
2475
-20° C, Avoid Freeze/Thaw Cycles