missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTOR Monoclonal Antibody (OTI5B3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805914
This item is not returnable.
View return policy
Description
Mammalian target of rapamycin (mTOR) is a serine/threonine kinase that plays a key role in cell growth, cell proliferation, and protein synthesis. mTOR mediates phosphoinositide 3-kinase and Akt/PKB signaling, resulting in phosphorylation of 4EBP1, and initiation of mRNA translation. A second pathway involves regulation of ribosomal S6 kinase, which affects ribosome biogenesis and translation elongation. Mutations affecting the gene results in Smith-Kingsmore syndrome.Specifications
| MTOR | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P42345 | |
| MTOR | |
| Human recombit protein fragment corresponding to amino acids 1766-2144 of human MTOR produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5B3 | |
| Unconjugated | |
| MTOR | |
| FRAP, FRAP1, FRAP2, RAFT1, RAPT1, SKS | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2475 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction