Learn More
MTOR Mouse anti-Human, Clone: OTI4A6, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF806072
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Mammalian target of rapamycin (mTOR) is a serine/threonine kinase that plays a key role in cell growth, cell proliferation, and protein synthesis. mTOR mediates phosphoinositide 3-kinase and Akt/PKB signaling, resulting in phosphorylation of 4EBP1, and initiation of mRNA translation. A second pathway involves regulation of ribosomal S6 kinase, which affects ribosome biogenesis and translation elongation. Mutations affecting the gene results in Smith-Kingsmore syndrome.Specifications
| MTOR | |
| Monoclonal | |
| Unconjugated | |
| MTOR | |
| FRAP, FRAP1, FRAP2, RAFT1, RAPT1, SKS | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2475 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4A6 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P42345 | |
| MTOR | |
| Human recombit protein fragment corresponding to amino acids 1766-2144 of human MTOR produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.