Learn More
mTOR Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-28273-100UL
Description
The mTOR antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human mTOR. This antibody recognizes human antigen. The mTOR antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Specifications
| mTOR | |
| 1000 μg/mL | |
| FLJ44809, FRAP, FRAP1, FRAP2, Mammalian target of rapamycin, MTOR, RAFT1, Rapamycin target protein 1, RAPT1 | |
| 100 μL | |
| 2475 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |