Learn More
mTOR Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-28273-100UL
 View more versions of this product
Product Code. 18049445
  • € 450.00 / 100µL
Description
The mTOR antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human mTOR. This antibody recognizes human antigen. The mTOR antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
mTOR
1000 μg/mL
FLJ44809, FRAP, FRAP1, FRAP2, Mammalian target of rapamycin, MTOR, RAFT1, Rapamycin target protein 1, RAPT1
100 μL
2475
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence, Flow Cytometry (Intracellular Staining)
CoraLite Plus 488
Rabbit
Primary
Human
150 kDa