missing translation for 'onlineSavingsMsg'
Learn More

mTOR Rabbit Recombinant Antibody, Proteintech

Product Code. 30233355 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30233355 20 μL 20µL
30237237 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30233355 Supplier Proteintech Supplier No. 816701RR20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Antibody

The mTOR antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human mTOR. This antibody recognizes human, mouse, rat antigen. The mTOR antibody has been validated for the following applications: WB, IHC, IF/ICC, FC (Intra), IP, ELISA analysis.

Specifications

Antigen mTOR
Applications Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining), Immunoprecipitation, ELISA
Classification Recombinant Monoclonal
Clone 6H23
Concentration 300 μg/mL
Conjugate Unconjugated
Gene Symbols FLJ44809; FRAP; FRAP2; MTOR; RAFT1; RAPT1
Host Species Rabbit
Immunogen Recombinant Protein
Purification Method Protein A purification
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2475
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 250-289 kDa
Product Type Recombinant Antibody
Form Liquid
Isotype IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.