missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTP18 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93000-0.1ml
This item is not returnable.
View return policy
Description
MTP18 Polyclonal antibody specifically detects MTP18 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| MTP18 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| HSPC242, Mitochondrial 18 kDa protein, mitochondrial fission process 1, mitochondrial fission process protein 1, mitochondrial protein 18 kDa, MTP18mitochondrial fission protein MTP18 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-166 of human MTFP1 (NP_057582.2). MSEPQPRGAERDLYRDTWVRYLGYANEVGEAFRSLVPAAVVWLSYGVASSYVLADAIDKGKKAGEVPSPEAGRSARVTVAVVDTFVWQALASVAIPGFTINRVCAASLYVLGTATRWPLAVRKWTTTALGLLTIPIIIHPIDRSVDFLLDSSLRKLYPTVGKPSSS | |
| 0.1 mL | |
| Apoptosis | |
| 51537 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction