missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTRF1L Antibody (OTI1D11), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72814H
This item is not returnable.
View return policy
Description
MTRF1L Monoclonal specifically detects MTRF1L in Human samples. It is validated for Western Blot.
Specifications
| MTRF1L | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| MTRF1L | |
| Full length recombinant protein of human MTRF1L (NP_061914) produced in HEK293T cells. | |
| 0.1 mL | |
| 54516.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI1D11 | |
| Western Blot | |
| mitochondrial translational release factor 1-likemitochondrial, MRF1L, MTRF1A | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?