missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTRF1L Antibody (OTI1D11), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72814MFV500
This item is not returnable.
View return policy
Description
MTRF1L Monoclonal antibody specifically detects MTRF1L in Human samples. It is validated for Flow Cytometry
Specifications
| MTRF1L | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 54516 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| OTI1D11 | |
| Flow Cytometry | |
| mitochondrial translational release factor 1-likemitochondrial, MRF1L, MTRF1A | |
| Full length human recombinant protein of human MTRF1L (NP_061914) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction