missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTRF1L Monoclonal Antibody (OTI1H7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501003S
This item is not returnable.
View return policy
Description
MTRF1L Monoclonal antibody specifically detects MTRF1L in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| MTRF1L | |
| Monoclonal | |
| 0.67 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y5X9 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9388 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1H7 | |
| Unconjugated | |
| LIPG | |
| 9130004K12Rik; HMRF1L; mitochondrial release factor 1 like; mitochondrial translational release factor 1 like; mitochondrial translational release factor 1-like; MRF1L; mtRF1a; MTRF1L; Peptide chain release factor 1-like, mitochondrial | |
| Full length human recombinant protein of human LIPG produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction