Learn More
MTRF1L Monoclonal Antibody (OTI1H5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA501068
Encompass_Preferred
 View more versions of this product
Product Code. 17523201
  • € 540.87 / 100µL
Description
Mitochondrial peptide chain release factor that directs the termination of translation in response to the peptide chain termination codons UAA and UAG.
Specifications
Specifications
MTRF1L
Monoclonal
0.67 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9UGC7
MTRF1L
Full length human recombit protein of human MTRF1L produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI1H5
Unconjugated
MTRF1L
HMRF1L, MRF1L, mtRF1a
Mouse
Affinity Chromatography
RUO
54516
-20° C, Avoid Freeze/Thaw Cycles