missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mtTFA Antibody (18G102B2E11) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80858
This item is not returnable.
View return policy
Description
mtTFA Monoclonal specifically detects mtTFA in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated, Knockdown Validated.
Specifications
| mtTFA | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Simple Western : 1:20, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence : 1:50, Immunohistochemistry-Paraffin : 1:200, Knockout Validated, KnockDown Validated | |
| mitochondrial transcription factor A, MtTF1, mtTFA, TCF-6, TCF6L1, TCF6L2Mitochondrial transcription factor 1, TCF6TCF6L3, Transcription factor 6, Transcription factor 6-like 2, Transcription factor 6-like 2 (mitochondrial transcription factor), transcription factor A, mitochondrial | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human and rat. Mouse reactivity reported in scientific literature (PMID: 28053692) | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay, KnockDown | |
| 18G102B2E11 | |
| Unconjugated | |
| Tris-Glycine, 0.15 M NaCl | |
| TFAM | |
| Human mtTFA [Swiss-Prot# Q00059] | |
| 0.1 mL | |
| Cell Cycle and Replication, Transcription Factors and Regulators | |
| 7019 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction