missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119279
This item is not returnable.
View return policy
Description
MTX1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
MTX1 belongs to the metaxin family. It is involved in transport of proteins into the mitochondrion and essential for embryonic development.
Specifications
| MTX1 | |
| Polyclonal | |
| Unconjugated | |
| MTX1 | |
| Gcap6; LOW QUALITY PROTEIN: metaxin-1; Metaxin 1; metaxin-1; Mitochondrial outer membrane import complex protein 1; Mtx; MTX1; MTXN; Unknown (protein for MGC:134321) | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 295241 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| 0 | |
| MTX1 | |
| Recombinant protein Metaxin 1. | |
| 200 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction