missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTX1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121761
This item is not returnable.
View return policy
Description
MTX1 Polyclonal Antibody for Western Blot, IHC (P)
MTX1 belongs to the metaxin family. It is involved in transport of proteins into the mitochondrion and essential for embryonic development.
Specifications
| MTX1 | |
| Polyclonal | |
| Unconjugated | |
| MTX1 | |
| Gcap6; LOW QUALITY PROTEIN: metaxin-1; Metaxin 1; metaxin-1; Mitochondrial outer membrane import complex protein 1; Mtx; MTX1; MTXN; Unknown (protein for MGC:134321) | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 295241, 4580 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q13505 | |
| MTX1 | |
| Recombinant protein Metaxin 1. The antigen corresponds to amino acid range 175-387 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction