missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTX3 Antibody (S08-2K4), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 421.00 - € 664.00
Specifications
| Antigen | MTX3 |
|---|---|
| Clone | S08-2K4 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18699393
|
Novus Biologicals
NBP3-19703-100ul |
100 μg |
€ 664.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18696853
|
Novus Biologicals
NBP3-19703-50ul |
50 μg |
€ 421.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MTX3 Monoclonal antibody specifically detects MTX3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| MTX3 | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 345778 | |
| IgG | |
| Affinity purified |
| S08-2K4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| MTX3 metaxin 3 | |
| A synthetic peptide of human MTX3 (Uniprot # Q5HYI7) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title