missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MU 1742
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
Description
MU 1742 is a potent and selective casein kinase 1α (CK1α), CK1δ and CK1ε inhibitor (IC50 values are 6, 7 and 28 nM for CK1δ, CK1α and CK1ε respectively). In vivo, MU 1742 inhibits CK1δ/ε-dependent DVL3 phosphorylation in mouse lung tissue. MU 1742 is orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 4-(1-((4-Fluoro-1-methylpiperidin-4-yl)methyl)-4-(5-fluoropyridin-2-yl)-1H-imidazol-5-yl)-1H-pyrrolo[2,3-b]pyridine |
| Quantity | 2 mg |
| Target | Casein Kinase 1 Inhibitors |
| Molecular Formula | C22H22F2N6 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction