missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mu Crystallin Antibody (OTI1G7), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72825AF350
This item is not returnable.
View return policy
Description
mu Crystallin Monoclonal antibody specifically detects mu Crystallin in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| mu Crystallin | |
| Monoclonal | |
| Alexa Fluor 350 | |
| crystallin, mu, DFNA40NADP-regulated thyroid-hormone binding protein, mu-crystallin homolog, NADP-regulated thyroid-hormone-binding protein, THBP | |
| Full length recombinant protein of human mu Crystallin (NP_001879) produced in HEK293T cells. | |
| 0.1 mL | |
| Vision | |
| 1428 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI1G7 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction