missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC1 Antibody (2336A), DyLight 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB62981M
This item is not returnable.
View return policy
Description
MUC1 Monoclonal antibody specifically detects MUC1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry
Specifications
| MUC1 | |
| Monoclonal | |
| DyLight 594 | |
| Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin | |
| Chinese hamster ovary cell line CHO-derived recombinant human MUC-1, Met1-Ser380, Accession # P15941 | |
| 0.1 mL | |
| Cancer, Cellular Markers, Extracellular Matrix, Inflammation, Signal Transduction | |
| 4582 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry, Immunohistochemistry | |
| 2336A | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur