Learn More
MUC1 Antibody (604804), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB6298MFV500
Product Code. 18551546
  • € 684.00 / 0.10mL
Description
MUC1 Monoclonal antibody specifically detects MUC1 in Human samples. It is validated for Western Blot, Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MUC1
Monoclonal
mFluor Violet 500 SE
50mM Sodium Borate
Mouse
0.1 mL
Cancer, Cellular Markers, Extracellular Matrix, Inflammation, Signal Transduction
4582
Store at 4C in the dark.
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry
604804
Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready
Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin
E. coli-derived recombinant human MUC-1, Pro126-Arg145, Accession # P15941
RUO
Primary
Human
IgG2b