missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NBP1-60046
This item is not returnable.
View return policy
Description
MUC1 Polyclonal specifically detects MUC1 in Human, Mouse, Rat, Porcine, Bovine, Equine, Guinea Pig, Goat, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| MUC-1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin | |
| Rabbit | |
| 21 kDa | |
| 0.1 mg | |
| Cancer, Cellular Markers, Extracellular Matrix, Inflammation, Signal Transduction | |
| 4582 | |
| Human, Mouse, Rat, Pig, Bovine, Equine, Guinea Pig, Goat, Rabbit | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 0.2-1 ug/ml, Flow Cytometry reported in scientific literature (PMID 28740504), Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 4-8 ug/ml, Knockout Validated | |
| Q7Z552 | |
| MUC1 | |
| Synthetic peptides corresponding to MUC1(mucin 1, cell surface associated) The peptide sequence was selected from the C terminal of MUC1 [NP_001037855]. Peptide sequence GQLDIFPARDTYHPMSEYPTYHTHGRYVPPSSTDRSPYEKVSAGNGGSSL. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Guinea pig: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Bovine: 92%. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction