missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC1 Antibody (HMPV), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47883PECY55
This item is not returnable.
View return policy
Description
MUC1 Monoclonal specifically detects MUC1 in Human samples. It is validated for Flow Cytometry.
Specifications
| MUC1 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| MUC1 | |
| Human breast cancer cell line ZR-75 cells | |
| 0.1 mL | |
| Primary | |
| This monoclonal antibody recognizes full-length MUC1 in a glycosylation-independent manner and can bind to the fully glycosylated protein. The dominant epitope of this monoclonal antibody is APDTR in the VNTR region. It reacts with the core peptide of the MUC1 protein, which is a member of a family of mucin glycoproteins that are characterized by high carbohydrate content, O-linked oligosaccharides, high molecular weight (200kDa) and an amino acid composition rich in serine, threonine, proline and glycine. The core protein contains a domain of 20 amino-acid tandem repeats that functions as multiple epitopes for the monoclonal antibody. Incomplete glycosylation of some tumor-associated mucins may lead to variable unmasking of the multiple peptide epitopes leading to the observed differences in staining intensity between normal and malignant tissues. This monoclonal antibody reacts with both normal and malignant epithelia of various tissues including breast and colon. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| HMPV | |
| Flow Cytometry | |
| Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin | |
| Mouse | |
| Protein A or G purified | |
| Cancer, Cellular Markers, Extracellular Matrix, Inflammation, Signal Transduction | |
| 4582.0 | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction