missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC1 Antibody (MUC1/520), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47886AF350
This item is not returnable.
View return policy
Description
MUC1 Monoclonal specifically detects MUC1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| MUC1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| MUC1 | |
| Human milk-fat globule membranes (HMFGM) (Uniprot: P15941 ) | |
| 0.1 mL | |
| Primary | |
| In Western blotting, it recognizes proteins in MW range of 265-400kDa, identified as different glycoforms of EMA. EMA may provide a protective layer on epithelial cells against bacterial and enzyme attack. In immunohistochemical assays, it superbly stains routine formalin/paraffin carcinomas. Antibody to EMA is useful as a pan-epithelial marker for detecting early metastatic loci of carcinoma in bone marrow or liver. | |
| Store at 4°C in the dark. |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| MUC1/520 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence | |
| Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin | |
| Mouse | |
| Protein A or G purified | |
| Cancer, Cellular Markers, Extracellular Matrix, Inflammation, Signal Transduction | |
| 4582.0 | |
| Human | |
| IgG2a λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction