missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC1 Antibody (MUC1/7797R), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 268.00 - € 496.00
Specifications
| Antigen | MUC1 |
|---|---|
| Clone | MUC1/7797R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18671565
|
Novus Biologicals
NBP3-23889-20ug |
20 μg |
€ 268.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18694984
|
Novus Biologicals
NBP3-23889-100ug |
100 μg |
€ 496.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MUC1 Monoclonal antibody specifically detects MUC1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| MUC1 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cellular Markers, Extracellular Matrix, Inflammation, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| 4582 | |
| IgG κ | |
| Protein A or G purified |
| MUC1/7797R | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin | |
| Recombinant full-length human MUC1 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title