missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC1 Antibody (MUC1/967), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47885AF488
This item is not returnable.
View return policy
Description
MUC1 Monoclonal antibody specifically detects MUC1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
Specifications
| MUC1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin | |
| Human milk-fat globule membranes (HMFGM) (Uniprot: P15941 ) | |
| 0.1 mL | |
| Cancer, Cellular Markers, Extracellular Matrix, Inflammation, Signal Transduction | |
| 4582 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| MUC1/967 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction