missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC1 Antibody (OTI2E3), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72826AF647
This item is not returnable.
View return policy
Description
MUC1 Monoclonal specifically detects MUC1 in Human, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| MUC1 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MUC1 | |
| Full length recombinant protein of human MUC-1 (NP_001018016) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Canine, Monkey | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| OTI2E3 | |
| Western Blot, Immunohistochemistry | |
| Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin | |
| Mouse | |
| Affinity Purified | |
| Cancer, Cellular Markers, Extracellular Matrix, Inflammation, Signal Transduction | |
| 4582.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction