missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MUC13 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142343
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:4,000.
Mucin 13, epithelial transmembrane. SEA is an extracellular domain associated with O-glycosylation. Proteins found to contain SEA-modules include, agrin, enterokinase, 63 kDa sea urchin (Strongylocentrotus purpuratus) sperm protein, perlecan (heparan sulphate proteoglycan core, mucin 1 and the cell surface antigen, 114/A10, and two functionally uncharacterised, probably extracellular, Caenorhabditis elegans proteins. Despite the functional diversity of these adhesive proteins, a common denominator seems to be their existence in heavily glycosylated environments. In addition, the better characterised proteins all contain O-glycosidic-linked carbohydrates such as heparan sulphate that contribute considerably to their molecular masses. The common module might regulate or assist binding to neighbouring carbohydrate moieties. Enterokinase, the initiator of intestinal digestion, is a mosaic protease composed of a distinctive assortment of domains.
Specifications
| MUC13 | |
| Polyclonal | |
| Unconjugated | |
| MUC13 | |
| 114/A10; 14/A10; AI159736; Cell surface antigen 114/A10; down-regulated in colon cancer 1; DRCC1; Lrrp; Ly64; Lymphocyte antigen 64; Muc-123; MUC13; MUC-13; mucin 13; mucin 13, cell surface associated; mucin 13, epithelial transmembrane; mucin-13; NJ-1; RECC; UNQ6194/PRO20221 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 56667 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9H3R2 | |
| MUC13 | |
| Peptide with sequence DCKSDLQRPNPQ. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction