Learn More
Novus Biologicals™ MUC17 Antibody (Forerunner patent anti-Muc 17) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28802-50ug
 View more versions of this product
Product Code. 30047234
  • € 213.00 / 50µg
Description
MUC17 Monoclonal antibody specifically detects MUC17 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MUC17
Monoclonal
Unconjugated
Q685J3
Human
Protein A purified
RUO
140453
Human
Purified
Flow Cytometry, ELISA, Functional Assay
Forerunner patent anti-Muc 17
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
membrane mucin MUC17, MUC-3, mucin 17, cell surface associated, secreted mucin MUC17, small intestinal mucin MUC3, small intestinal mucin-3
MUC17
50 μg
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.