missing translation for 'onlineSavingsMsg'
Learn More

MUC17 Antibody (Forerunner patent anti-Muc 17) [Janelia Fluor™ 585], Novus Biologicals™

Product Code. 30170531
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30170531 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30170531 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP328802JF585

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

MUC17 Monoclonal antibody specifically detects MUC17 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen MUC17
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone Forerunner patent anti-Muc 17
Conjugate Janelia Fluor 585
Formulation 50mM Sodium Borate
Gene Alias membrane mucin MUC17, MUC-3, mucin 17, cell surface associated, secreted mucin MUC17, small intestinal mucin MUC3, small intestinal mucin-3
Host Species Human
Immunogen MUC17
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140453
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.