missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC2 Antibody (994/152), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50521AF647
This item is not returnable.
View return policy
Description
MUC2 Monoclonal specifically detects MUC2 in Human samples. It is validated for ELISA, Immunohistochemistry.
Specifications
| MUC2 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MUC2 | |
| This MUC2 Antibody (994/152) was developed against a peptide based on the tandem repeat sequence of MUC2 | |
| 0.1 mL | |
| Cancer, Cellular Markers, Inflammation, Signal Transduction | |
| 4583 | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry | |
| 994/152 | |
| ELISA, Immunohistochemistry | |
| Intestinal mucin-2, MLP, MUC-2, mucin 2, intestinal/tracheal, mucin 2, oligomeric mucus/gel-forming, mucin-2, mucin-like protein, SMUC | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| MUC2 Antibody (994/152) is specific for Muc2 and will not detect Muc3 or Muc4. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction