missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC2 Antibody (994/152), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50521DL594
This item is not returnable.
View return policy
Description
MUC2 Monoclonal antibody specifically detects MUC2 in Human samples. It is validated for ELISA, Immunohistochemistry
Specifications
| MUC2 | |
| Monoclonal | |
| DyLight 594 | |
| Intestinal mucin-2, MLP, MUC2, MUC-2, mucin 2, intestinal/tracheal, mucin 2, oligomeric mucus/gel-forming, mucin-2, SMUC | |
| This MUC2 Antibody (994/152) was developed against a peptide based on the tandem repeat sequence of MUC2 | |
| 0.1 mL | |
| Cancer, Cellular Markers, Inflammation, Signal Transduction | |
| 4583 | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA, Immunohistochemistry | |
| 994/152 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur