missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MUC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
€ 157.00 - € 442.00
Specifications
| Antigen | MUC2 |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17770694
![]() |
Invitrogen™
PA5119291 |
200 μL |
€ 442.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17763154
|
Invitrogen™
PA5121758 |
20 μL |
€ 157.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MUC2 Polyclonal Antibody for Western Blot, IHC (P)
Secreted epithelial mucins are large macromolecules which exhibit extreme polydispersity. Mucin 2 is the major intestinal mucin. O-glycans are attached to MUC2 in a potentially diverse arrangement, which is crucial for their interaction with endogeneous and exogeneous lectins.Specifications
| MUC2 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Mouse | |
| MUC2 | |
| 2010015E03Rik; colonic mucin; HH-Muc; intestinal mucin-2; MCM; MLP; muc2; MUC-2; mucin 2; mucin 2, intestinal/tracheal; mucin 2, oligomeric mucus/gel-forming; mucin-2; mucin-like protein; secreted gel-forming mucin; SMUC; wnn | |
| MUC2 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q80Z19 | |
| 17831 | |
| Recombinant protein Mucin 2. The antigen corresponds to amino acid range 27-110 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title