missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC2 Rabbit anti-Human, Clone: MLP/2970R, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-07238-100ug
This item is not returnable.
View return policy
Description
MUC2 Monoclonal specifically detects MUC2 in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications
| MUC2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Intestinal mucin-2, MLP, MUC2, MUC-2, mucin 2, intestinal/tracheal, mucin 2, oligomeric mucus/gel-forming, mucin-2, SMUC | |
| Recombinant full-length human MUC2 protein (exact sequence is proprietary) | |
| 100 μg | |
| Cancer, Cellular Markers, Inflammation, Signal Transduction | |
| 4583 | |
| Store at 4°C. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| MLP/2970R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction