missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC20 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94455-0.1ml
This item is not returnable.
View return policy
Description
MUC20 Polyclonal antibody specifically detects MUC20 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| MUC20 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FLJ14408, FLJ53153, KIAA1359mucin-20, MUC-20, mucin 20, cell surface associated, transmembrane mucin MUC20S | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human MUC20 (NP_689886.3). PNFMVLIATSVETSAASGSPEGAGMTTVQTITGSDPEEAIFDTLCTDDSSEEAKTLTMDILTLAHTSTEAKGLSSESSASSDGPHPVITPSRASESSASSD | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 200958 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction