missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC3 Antibody (SPM200), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-11579CL1
This item is not returnable.
View return policy
Description
MUC3 Monoclonal antibody specifically detects MUC3 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Paraffin),Immunofluorescence,Immunocytochemistry/Immunofluorescence
Specifications
| MUC3 | |
| Monoclonal | |
| CoraFluor 1 | |
| MUC-3A, mucin 3, intestinal, mucin 3A, cell surface associated, mucin 3A, intestinal, mucin-3A | |
| A synthetic peptide of 35 amino acids, SIB35 (C-HSTPSFTSSITTTETTSHSTPSFTSSITTTETTS), which contains two of the MUC3 tandem repeats, coupled to KLH. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry/Immunofluorescence | |
| SPM200 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 4584 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction