missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC5AC Antibody (2-11M1), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11418AF350
This item is not returnable.
View return policy
Description
MUC5AC Monoclonal antibody specifically detects MUC5AC in Human, Mouse, Bovine, Feline, Monkey samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
Specifications
| MUC5AC | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Bovine, Feline, Monkey | |
| Purified |
| Flow Cytometry, ELISA, Immunofluorescence | |
| 2-11M1 | |
| Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| gastric mucin, leB, lewis B blood group antigen, major airway glycoprotein, MUC5, mucin 5, subtypes A and C, tracheobronchial/gastric, mucin 5AC, oligomeric mucus/gel-forming, mucin 5AC, oligomeric mucus/gel-forming pseudogene, mucin-5 subtype AC, tracheobronchial, mucin-5AC, TBM, tracheobronchial mucin | |
| M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b) patient | |
| 0.1 mL | |
| Cellular Markers, Extracellular Matrix, Infections (Virus Bacteria and Parasites), Signal Transduction | |
| 4586 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction